4-(2-Aminoacetyl)-2-piperazinone hydrochloride

Supplier: Matrix Scientific

052204-500MG 052204-2.5G
101898-702EA 364.62 USD
101898-702 101898-704
4-(2-Aminoacetyl)-2-piperazinone hydrochloride
4-(2-Aminoacetyl)-2-piperazinone hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClN₃O₂
MDL Number: MFCD13561982
CAS Number: 1220017-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR