N-Benzyl-5-bromo-N-ethyl-2-pyridinamine
Supplier: Matrix Scientific
052158-2.5G
052158-500MG
101898-522EA
364.62
USD
101898-522
101898-524
N-Benzyl-5-bromo-N-ethyl-2-pyridinamine
N-Benzyl-5-bromo-N-ethyl-2-pyridinamine
Hazard: Irritant
Formula:
C₁₄H₁₅BrN₂ |
MDL Number:
MFCD13476774 CAS Number: 1220030-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...