1-(5-Bromo-2-pyridinyl)indoline

Supplier: Matrix Scientific

052156-500MG 052156-2.5G
101898-514EA 364.62 USD
101898-514 101898-516
1-(5-Bromo-2-pyridinyl)indoline
1-(5-Bromo-2-pyridinyl)indoline

Hazard: Irritant

Formula: C₁₃H₁₁BrN₂
MDL Number: MFCD13561947
CAS Number: 1220033-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR