N-(5-Bromo-3-methyl-2-pyridinyl)-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)amine
Supplier: Matrix Scientific
052130-2.5G
052130-500MG
101898-410EA
364.62
USD
101898-410
101898-412
N-(5-Bromo-3-methyl-2-pyridinyl)-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)amine
N-(5-Bromo-3-methyl-2-pyridinyl)-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)amine
Hazard: Irritant
Formula:
C₁₃H₁₉BrN₂O |
MDL Number:
MFCD13561936 CAS Number: 1220036-22-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...