N-Benzyl-5-bromo-N-ethyl-3-methyl-2-pyridinamine

Supplier: Matrix Scientific

052129-500MG 052129-2.5G
101898-406EA 364.62 USD
101898-406 101898-408
N-Benzyl-5-bromo-N-ethyl-3-methyl-2-pyridinamine
N-Benzyl-5-bromo-N-ethyl-3-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₅H₁₇BrN₂
MDL Number: MFCD13561935
CAS Number: 1219971-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR