1-(5-Bromo-3-methyl-2-pyridinyl)indoline

Supplier: Matrix Scientific

052127-500MG 052127-2.5G
101898-398EA 364.62 USD
101898-398 101898-400
1-(5-Bromo-3-methyl-2-pyridinyl)indoline
1-(5-Bromo-3-methyl-2-pyridinyl)indoline

Hazard: Irritant

Formula: C₁₄H₁₃BrN₂
MDL Number: MFCD13561933
CAS Number: 1220016-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR