1-(5-Bromo-3-methyl-2-pyridinyl)azepane
Supplier: Matrix Scientific
052117-2.5G
052117-500MG
101898-358EA
364.62
USD
101898-358
101898-360
1-(5-Bromo-3-methyl-2-pyridinyl)azepane
1-(5-Bromo-3-methyl-2-pyridinyl)azepane
Hazard: Irritant
Formula:
C₁₂H₁₇BrN₂ |
MDL Number:
MFCD13561923 CAS Number: 1220017-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...