5-Bromo-3-methyl-2-(4-methyl-1-piperidinyl)pyridine
Supplier: Matrix Scientific
052114-2.5G
052114-500MG
101898-346EA
364.62
USD
101898-346
101898-348
5-Bromo-3-methyl-2-(4-methyl-1-piperidinyl)pyridine
5-Bromo-3-methyl-2-(4-methyl-1-piperidinyl)pyridine
Hazard: Irritant
Formula:
C₁₂H₁₇BrN₂ MW: 269.19 g/mol |
MDL Number:
MFCD13561920 CAS Number: 1220034-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...