5-Bromo-3-methyl-2-(1-pyrrolidinyl)pyridine

Supplier: Matrix Scientific

052103-500MG 052103-2.5G
101898-302EA 364.62 USD
101898-302 101898-304
5-Bromo-3-methyl-2-(1-pyrrolidinyl)pyridine
5-Bromo-3-methyl-2-(1-pyrrolidinyl)pyridine

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂
MDL Number: MFCD13561910
CAS Number: 1220016-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR