4-Chloro-6-[3,4-dihydro-2(1H)-isoquinolinyl]-1,3,5-triazin-2-amine
Supplier: Matrix Scientific
052097-2.5G
052097-500MG
101898-278EA
364.62
USD
101898-278
101898-280
4-Chloro-6-[3,4-dihydro-2(1H)-isoquinolinyl]-1,3,5-triazin-2-amine
4-Chloro-6-[3,4-dihydro-2(1H)-isoquinolinyl]-1,3,5-triazin-2-amine
Hazard: Irritant
Formula:
C₁₂H₁₂ClN₅ |
MDL Number:
MFCD13561904 CAS Number: 1219971-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...