4-Chloro-6-(2-ethyl-1-piperidinyl)-1,3,5-triazin-2-amine

Supplier: Matrix Scientific

052093-500MG 052093-2.5G
101898-264EA 838.46 USD
101898-264 101898-262
4-Chloro-6-(2-ethyl-1-piperidinyl)-1,3,5-triazin-2-amine
4-Chloro-6-(2-ethyl-1-piperidinyl)-1,3,5-triazin-2-amine

Hazard: Irritant

Formula: C₁₀H₁₆ClN₅
MDL Number: MFCD13561900
CAS Number: 1220038-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR