4-(1-Azepanyl)-6-chloro-1,3,5-triazin-2-amine
Supplier: Matrix Scientific
052088-2.5G
052088-500MG
101898-242EA
364.62
USD
101898-242
101898-244
4-(1-Azepanyl)-6-chloro-1,3,5-triazin-2-amine
4-(1-Azepanyl)-6-chloro-1,3,5-triazin-2-amine
Hazard: Irritant
Formula:
C₉H₁₄ClN₅ |
MDL Number:
MFCD13561896 CAS Number: 1220031-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...