N-[5-(Aminomethyl)-2-pyridinyl]-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)amine

Supplier: Matrix Scientific

052073-500MG 052073-2.5G
101898-182EA 364.62 USD
101898-182 101898-184
N-[5-(Aminomethyl)-2-pyridinyl]-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)amine
N-[5-(Aminomethyl)-2-pyridinyl]-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)amine

Hazard: Irritant

Formula: C₁₃H₂₁N₃O
MDL Number: MFCD13561885
CAS Number: 1220016-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR