5-(Aminomethyl)-N-butyl-N-methyl-2-pyridinamine
Supplier: Matrix Scientific
052052-2.5G
052052-500MG
101898-100EA
838.46
USD
101898-100
101898-098
5-(Aminomethyl)-N-butyl-N-methyl-2-pyridinamine
5-(Aminomethyl)-N-butyl-N-methyl-2-pyridinamine
Hazard: Irritant
Formula:
C₁₁H₁₉N₃ |
MDL Number:
MFCD09811492 CAS Number: 1016510-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...