2-[[5-(Aminomethyl)-2-pyridinyl](methyl)amino]-1-ethanol

Supplier: Matrix Scientific

052048-500MG 052048-2.5G
101898-082EA 461.54 USD
101898-082 101898-084
2-[[5-(Aminomethyl)-2-pyridinyl](methyl)amino]-1-ethanol
2-[[5-(Aminomethyl)-2-pyridinyl](methyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₉H₁₅N₃O
MDL Number: MFCD12813428
CAS Number: 1178939-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR