(2-Chloro-3-pyridinyl)(2-ethyl-1-piperidinyl)methanone

Supplier: Matrix Scientific

052036-500MG 052036-2.5G
101898-034EA 364.62 USD
101898-034 101898-036
(2-Chloro-3-pyridinyl)(2-ethyl-1-piperidinyl)methanone
(2-Chloro-3-pyridinyl)(2-ethyl-1-piperidinyl)methanone

Hazard: Irritant

Formula: C₁₃H₁₇ClN₂O
MDL Number: MFCD01336311
CAS Number: 573993-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR