1-Azepanyl(2-chloro-3-pyridinyl)methanone
Supplier: Matrix Scientific
052031-2.5G
052031-500MG
101898-014EA
364.62
USD
101898-014
101898-016
1-Azepanyl(2-chloro-3-pyridinyl)methanone
1-Azepanyl(2-chloro-3-pyridinyl)methanone
Hazard: Irritant
Formula:
C₁₂H₁₅ClN₂O |
MDL Number:
MFCD02371363 CAS Number: 438467-27-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...