N2-Cyclohexyl-N2-methyl-2,3-pyridinediamine
Supplier: Matrix Scientific
052009-500MG
052009-2.5G
101897-928EA
838.46
USD
101897-928
101897-926
N2-Cyclohexyl-N2-methyl-2,3-pyridinediamine
N2-Cyclohexyl-N2-methyl-2,3-pyridinediamine
Hazard: Irritant
Formula:
C₁₂H₁₉N₃ |
MDL Number:
MFCD09737137 CAS Number: 954273-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...