2-[(3-Amino-2-pyridinyl)(ethyl)amino]-1-ethanol

Supplier: Matrix Scientific

051999-2.5G 051999-500MG
101897-888EA 1012.45 USD
101897-888 101897-886
2-[(3-Amino-2-pyridinyl)(ethyl)amino]-1-ethanol
2-[(3-Amino-2-pyridinyl)(ethyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₉H₁₅N₃O
MDL Number: MFCD11562398
CAS Number: 1082137-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR