2-[Benzyl(ethyl)amino]nicotinic acid
Supplier: Matrix Scientific
051990-2.5G
051990-500MG
101897-850EA
364.62
USD
101897-850
101897-852
2-[Benzyl(ethyl)amino]nicotinic acid
2-[Benzyl(ethyl)amino]nicotinic acid
Hazard: Irritant
Formula:
C₁₅H₁₆N₂O₂ |
MDL Number:
MFCD07379603 CAS Number: 1040076-82-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...