2-(3-Hydroxy-1-pyrrolidinyl)nicotinic acid
Supplier: Matrix Scientific
051970-2.5G
051970-500MG
101897-772EA
838.46
USD
101897-772
101897-770
2-(3-Hydroxy-1-pyrrolidinyl)nicotinic acid
2-(3-Hydroxy-1-pyrrolidinyl)nicotinic acid
Hazard: Irritant
Formula:
C₁₀H₁₂N₂O₃ |
MDL Number:
MFCD13561876 CAS Number: 1219968-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...