2-[Benzyl(ethyl)amino]isonicotinic acid
Supplier: Matrix Scientific
051964-500MG
051964-2.5G
101897-748EA
838.46
USD
101897-748
101897-746
2-[Benzyl(ethyl)amino]isonicotinic acid
2-[Benzyl(ethyl)amino]isonicotinic acid
Hazard: Irritant
Formula:
C₁₅H₁₆N₂O₂ |
MDL Number:
MFCD12463524 CAS Number: 1039979-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...