2-(4-Hydroxy-1-piperidinyl)isonicotinic acid
Supplier: Matrix Scientific
051954-2.5G
051954-500MG
101897-708EA
838.46
USD
101897-708
101897-706
2-(4-Hydroxy-1-piperidinyl)isonicotinic acid
2-(4-Hydroxy-1-piperidinyl)isonicotinic acid
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₃ |
MDL Number:
MFCD11132169 CAS Number: 167641-00-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...