2-(3-Oxo-1-piperazinyl)isonicotinic acid
Supplier: Matrix Scientific
051953-2.5G
051953-500MG
101897-704EA
838.46
USD
101897-704
101897-702
2-(3-Oxo-1-piperazinyl)isonicotinic acid
2-(3-Oxo-1-piperazinyl)isonicotinic acid
Hazard: Irritant
Formula:
C₁₀H₁₁N₃O₃ |
MDL Number:
MFCD11132185 CAS Number: 1019323-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...