N-[3-(Aminomethyl)-2-pyridinyl]-N-benzyl-N-ethylamine
Supplier: Matrix Scientific
051941-500MG
051941-2.5G
101897-656EA
838.46
USD
101897-656
101897-654
N-[3-(Aminomethyl)-2-pyridinyl]-N-benzyl-N-ethylamine
N-[3-(Aminomethyl)-2-pyridinyl]-N-benzyl-N-ethylamine
Hazard: Irritant
Formula:
C₁₅H₁₉N₃ |
MDL Number:
MFCD10692867 CAS Number: 1152523-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...