1-[3-(Aminomethyl)-2-pyridinyl]-4-piperidinol
Supplier: Matrix Scientific
051931-2.5G
051931-500MG
101897-616EA
838.46
USD
101897-616
101897-614
1-[3-(Aminomethyl)-2-pyridinyl]-4-piperidinol
1-[3-(Aminomethyl)-2-pyridinyl]-4-piperidinol
Hazard: Irritant
Formula:
C₁₁H₁₇N₃O |
MDL Number:
MFCD09737100 CAS Number: 954271-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...