[2-(4-Methyl-1-piperidinyl)-3-pyridinyl]methanamine
Supplier: Matrix Scientific
051926-2.5G
051926-500MG
101897-596EA
838.46
USD
101897-596
101897-594
[2-(4-Methyl-1-piperidinyl)-3-pyridinyl]methanamine
[2-(4-Methyl-1-piperidinyl)-3-pyridinyl]methanamine
Hazard: Irritant
Formula:
C₁₂H₁₉N₃ |
MDL Number:
MFCD09734331 CAS Number: 953727-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...