[2-(2-Methyl-1-piperidinyl)-3-pyridinyl]methanamine
Supplier: Matrix Scientific
051924-2.5G
051924-500MG
101897-586EA
364.62
USD
101897-586
101897-588
[2-(2-Methyl-1-piperidinyl)-3-pyridinyl]methanamine
[2-(2-Methyl-1-piperidinyl)-3-pyridinyl]methanamine
Hazard: Irritant
Formula:
C₁₂H₁₉N₃ |
MDL Number:
MFCD09735632 CAS Number: 953737-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...