[2-(1-Pyrrolidinyl)-3-pyridinyl]methanamine
Supplier: Matrix Scientific
051915-2.5G
051915-500MG
101897-552EA
838.46
USD
101897-552
101897-550
[2-(1-Pyrrolidinyl)-3-pyridinyl]methanamine
[2-(1-Pyrrolidinyl)-3-pyridinyl]methanamine
Hazard: Irritant
Formula:
C₁₀H₁₅N₃ MW: 177.25 g/mol |
MDL Number:
MFCD08690249 CAS Number: 859850-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...