1-Benzyl-4-(6-chloro-2-pyrazinyl)piperazine

Supplier: Matrix Scientific

051899-500MG 051899-2.5G
101897-488EA 838.46 USD
101897-488 101897-486
1-Benzyl-4-(6-chloro-2-pyrazinyl)piperazine
1-Benzyl-4-(6-chloro-2-pyrazinyl)piperazine

Hazard: Irritant

Formula: C₁₅H₁₇ClN₄
MDL Number: MFCD13561857
CAS Number: 1219967-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR