1-(6-Chloro-2-pyrazinyl)-4-methylpiperazine
Supplier: Matrix Scientific
Synonyms:
2-Chloro-6-(4-methylpiperazin-1-yl)pyrazine
051898-500MG
051898-2.5G
101897-484EA
479.52
USD
101897-484
101897-482
1-(6-Chloro-2-pyrazinyl)-4-methylpiperazine
1-(6-Chloro-2-pyrazinyl)-4-methylpiperazine
Hazard: Irritant
Formula:
C₉H₁₃ClN₄ MW: 212.68 g/mol |
MDL Number:
MFCD12024895 CAS Number: 61655-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...