2-[(6-Chloro-2-pyrazinyl)(methyl)amino]-1-ethanol
Supplier: Matrix Scientific
051888-2.5G
051888-500MG
101897-444EA
838.46
USD
101897-444
101897-442
2-[(6-Chloro-2-pyrazinyl)(methyl)amino]-1-ethanol
2-[(6-Chloro-2-pyrazinyl)(methyl)amino]-1-ethanol
Hazard: Irritant
Formula:
C₇H₁₀ClN₃O |
MDL Number:
MFCD13561851 CAS Number: 1219961-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...