2-Chloro-6-(1-pyrrolidinyl)pyrazine

Supplier: Matrix Scientific

051886-500MG 051886-2.5G
101897-436EA 1986.55 USD
101897-436 101897-434
2-Chloro-6-(1-pyrrolidinyl)pyrazine
2-Chloro-6-(1-pyrrolidinyl)pyrazine

Hazard: Irritant

Formula: C₈H₁₀ClN₃
MW: 183.64 g/mol
MDL Number: MFCD09864953
CAS Number: 1000339-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR