3-Chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

051884-500MG 051884-2.5G
101897-426EA 364.62 USD
101897-426 101897-428
3-Chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₃H₁₆ClF₃N₂O
MDL Number: MFCD13561850
CAS Number: 1220028-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR