1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1,2,3,4-tetrahydroquinoline
Supplier: Matrix Scientific
051882-2.5G
051882-500MG
101897-420EA
838.46
USD
101897-420
101897-418
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1,2,3,4-tetrahydroquinoline
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1,2,3,4-tetrahydroquinoline
Hazard: Irritant
Formula:
C₁₅H₁₂ClF₃N₂ |
MDL Number:
MFCD13561848 CAS Number: 1219967-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...