3-Chloro-N-cyclohexyl-N-ethyl-5-(trifluoromethyl)-2-pyridinamine
Supplier: Matrix Scientific
051878-500MG
051878-2.5G
101897-404EA
838.46
USD
101897-404
101897-402
3-Chloro-N-cyclohexyl-N-ethyl-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-cyclohexyl-N-ethyl-5-(trifluoromethyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₄H₁₈ClF₃N₂ |
MDL Number:
MFCD13561846 CAS Number: 1219967-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...