3-Chloro-N-cyclohexyl-N-methyl-5-(trifluoromethyl)-2-pyridinamine
Supplier: Matrix Scientific
051875-500MG
051875-2.5G
101897-392EA
838.46
USD
101897-392
101897-390
3-Chloro-N-cyclohexyl-N-methyl-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-cyclohexyl-N-methyl-5-(trifluoromethyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₃H₁₆ClF₃N₂ |
MDL Number:
MFCD13561843 CAS Number: 1220033-24-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...