4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-2-piperazinone
Supplier: Matrix Scientific
051873-2.5G
051873-500MG
101897-384EA
956.28
USD
101897-384
101897-382
4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-2-piperazinone
4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-2-piperazinone
Hazard: Irritant
Formula:
C₁₀H₉ClF₃N₃O |
MDL Number:
MFCD08876838 CAS Number: 923143-11-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...