1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-pyrrolidinol

Supplier: Matrix Scientific

051863-500MG 051863-2.5G
101897-344EA 270.77 USD
101897-344 101897-346
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-pyrrolidinol
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-pyrrolidinol

Hazard: Irritant

Formula: C₁₀H₁₀ClF₃N₂O
MW: 266.65 g/mol
MDL Number: MFCD13561836
CAS Number: 1219980-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR