N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-N,N-diethylamine
Supplier: Matrix Scientific
051860-2.5G
051860-500MG
101897-334EA
838.46
USD
101897-334
101897-332
N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-N,N-diethylamine
N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-N,N-diethylamine
Hazard: Irritant
Formula:
C₁₀H₁₂ClF₃N₂ |
MDL Number:
MFCD13561833 CAS Number: 1220029-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...