4-(4-Methyl-2-nitrophenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050780-2.5G 050780-500MG
101893-018EA 838.46 USD
101893-018 101893-016
4-(4-Methyl-2-nitrophenoxy)piperidine hydrochloride
4-(4-Methyl-2-nitrophenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₃
MDL Number: MFCD13561165
CAS Number: 1220034-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR