4-(tert-Pentyl)phenyl-3-piperidinyl ether hydrochloride
Supplier: Matrix Scientific
050760-500MG
050760-2.5G
101892-938EA
838.46
USD
101892-938
101892-936
4-(tert-Pentyl)phenyl-3-piperidinyl ether hydrochloride
4-(tert-Pentyl)phenyl-3-piperidinyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₆ClNO |
MDL Number:
MFCD13561147 CAS Number: 1219982-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...