4-[(4-Propoxyphenoxy)methyl]piperidine hydrochloride
Supplier: Matrix Scientific
050743-500MG
050743-2.5G
101892-872EA
364.62
USD
101892-872
101892-874
4-[(4-Propoxyphenoxy)methyl]piperidine hydrochloride
4-[(4-Propoxyphenoxy)methyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₄ClNO₂ |
MDL Number:
MFCD13561131 CAS Number: 1220037-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...