N-[4-(3-Piperidinyloxy)phenyl]acetamide hydrochloride

Supplier: Matrix Scientific

050735-500MG 050735-2.5G
101892-840EA 364.62 USD
101892-840 101892-842
N-[4-(3-Piperidinyloxy)phenyl]acetamide hydrochloride
N-[4-(3-Piperidinyloxy)phenyl]acetamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₂
MDL Number: MFCD13561123
CAS Number: 1220037-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR