4-(4-Chloro-3-methylphenoxy)piperidine hydrochloride
Supplier: Matrix Scientific
050726-2.5G
050726-500MG
101892-806EA
838.46
USD
101892-806
101892-804
4-(4-Chloro-3-methylphenoxy)piperidine hydrochloride
4-(4-Chloro-3-methylphenoxy)piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇Cl₂NO |
MDL Number:
MFCD13561114 CAS Number: 1220028-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...