Ethyl-3-(3-pyrrolidinyloxy)benzoate hydrochloride

050708-2.5G
101892-734EA 838.46 USD
101892-734
Ethyl-3-(3-pyrrolidinyloxy)benzoate hydrochloride
Ethyl-3-(3-pyrrolidinyloxy)benzoate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClNO₃
MDL Number: MFCD13561099
CAS Number: 1219960-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR