Ethyl-3-(4-piperidinylmethoxy)benzoate hydrochloride

050707-2.5G
101892-730EA 838.46 USD
101892-730
Ethyl-3-(4-piperidinylmethoxy)benzoate hydrochloride
Ethyl-3-(4-piperidinylmethoxy)benzoate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₃
MDL Number: MFCD13561098
CAS Number: 1219960-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR