Ethyl-3-(3-piperidinylmethoxy)benzoate hydrochloride

050706-2.5G
101892-726EA 838.46 USD
101892-726
Ethyl-3-(3-piperidinylmethoxy)benzoate hydrochloride
Ethyl-3-(3-piperidinylmethoxy)benzoate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₃
MDL Number: MFCD13561097
CAS Number: 1220027-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR