Ethyl-3-(3-piperidinyloxy)benzoate hydrochloride

050704-2.5G
101892-718EA 838.46 USD
101892-718
Ethyl-3-(3-piperidinyloxy)benzoate hydrochloride
Ethyl-3-(3-piperidinyloxy)benzoate hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClNO₃
MDL Number: MFCD13561095
CAS Number: 1220033-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR