3-(4-Bromo-2-isopropylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050377-500MG 050377-2.5G
101891-410EA 838.46 USD
101891-410 101891-408
3-(4-Bromo-2-isopropylphenoxy)piperidine hydrochloride
3-(4-Bromo-2-isopropylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560778
CAS Number: 1220032-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR